Ethyl octanoate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Caprylic acid ethyl ester, Octanoic acid ethyl ester, Ethyl caprylate
218825-100G
218825-500G
BT218825-100GEA
21
USD
BT218825-100G
BT218825-500G
Ethyl octanoate ≥99%
Ethyl octanoate
CAS: 106-32-1; EC No: 203-385-5; MDL No: MFCD00009552; RTECS: RH0680000
Liquid; Linear Formula: CH3(CH2)6COOCH2CH3; Molecular Formula: C10H20O2; MW: 172.26
Melting Point: -48°; Boiling Point: 206-208°; Flash point: 75°C (167°F)
Density (g/mL): 0.867; Refractive Index: 1.417
- RTECS: RH0680000
- Refractive index: 1.417
- Liquid
Formula:
C₁₀H₂₀O₂ Boiling Pt: 206…208 °C Melting Pt: –48 °C Density: 0.867 g/mL Flash Pt: 75 °C (167 °F) |
MDL Number:
MFCD00009552 CAS Number: 106-32-1 EINECS: 203-385-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...