6-Bromo-4-chloroquinoline ≥97%
Supplier: BeanTown Chemical
218805-1G
218805-250MG
BT218805-1GEA
447
USD
BT218805-1G
BT218805-250MG
6-Bromo-4-chloroquinoline ≥97%
6-Bromo-4-chloroquinoline
CAS: 65340-70-7; MDL No: MFCD00511001
Solid; Molecular Formula: C9H5BrClN ; MW: 242.5
- Solid
Formula:
C₉H₅BrClN MW: 242.50 g/mol |
MDL Number:
MFCD00511001 CAS Number: 65340-70-7 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...