Nickel ≥99.5% (trace metals basis), wire Ø 1.0 mm, hard type
Supplier: BeanTown Chemical
218755-10M
218755-50M
BT218755-10MEA
85
USD
BT218755-10M
BT218755-50M
Nickel ≥99.5% (trace metals basis), wire Ø 1.0 mm, hard type
Nickel
CAS: 7440-02-0; EC No: 231-111-4; MDL No: MFCD00011137
Solid; Molecular Formula: Ni; MW: 58.69
Melting Point: 1453°; Boiling Point: 2732°
Density (g/mL): 8.9
- Solid
Formula:
Ni Boiling Pt: 2732 °C Melting Pt: 1453 °C Density: 8.9 g/mL |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,III |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...