Adenosine 5'-monophosphate sodium salt
Supplier: BeanTown Chemical
Synonyms:
((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate xsodium salt
218750-1G
218750-5G
BT218750-1GEA
108.47
USD
BT218750-1G
BT218750-5G
Adenosine 5'-monophosphate sodium salt
Adenosine 5'-monophosphate sodium salt
CAS: 13474-03-8; EC No: 224-961-2; MDL No: MFCD08689634; RTECS: AU7481600
Powder; Molecular Formula: C10H13N5NaO7P; MW: 369.20
- RTECS: AU7481600
- Powder
Formula:
C₁₀H₁₄N₅NaO₇P MW: 370.214 g/mol |
MDL Number:
MFCD08689634 CAS Number: 13474-03-8 EINECS: 224-961-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...