2-Bromo-p-xylene ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Bromo-1,4-dimethylbenzene, 2,5-Dimethylbromobenzene

218705-100G 218705-25G 218705-500G
BT218705-100GEA 179 USD
BT218705-100G BT218705-25G BT218705-500G
2-Bromo-p-xylene ≥98%
2-Bromo-p-xylene

CAS: 553-94-6; EC No: 209-055-7; MDL No: MFCD00000074 Liquid; Molecular Formula: C8H9Br ; MW: 185.06 Melting Point: 9-10°; Boiling Point: 199-201°; Flash point: 79°C (174°F) Density (g/mL): 1.34; Refractive Index: 1.550


  • Refractive index: 1.550
  • Liquid

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 199…201 °C
Melting Pt: 9…10 °C
Density: 1.34 g/mL
Flash Pt: 79 °C (174 °F)
MDL Number: MFCD00000074
CAS Number: 553-94-6
EINECS: 209-055-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR