Aluminium oxide, α-phase ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
ɑ-Tonerde, α-Alumina
218655-100G
218655-2KG
218655-500G
BT218655-100GEA
32
USD
BT218655-100G
BT218655-2KG
BT218655-500G
Aluminium oxide, α-phase ≥99.9% (trace metals basis)
Aluminium oxide, α-phase
CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424
- <1.0 Micron APS Powder, S.A. 6-8 m²/g
Formula:
Al₂O₃ MW: 101.96 g/mol Melting Pt: 2040 °C |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...