EDTA ferric monosodium salt
Supplier: BeanTown Chemical
Synonyms:
Ethylenedinitrilotetra-acetic acid iron (III) sodium salt, Edetic acid iron (III) monosodium salt, Edetic acid iron (III) sodium salt, Ethylenediaminetetra-acetic acid iron (III) sodium salt, Iron (III) sodium ethylenediaminetetraacetate, Ethylenedinitrilotetra-acetic acid iron (III) monosodium salt, EDTA iron sodium, Sodium ferric ethylenediaminetetraacetate, Iron (III) monosodium ethylenediaminetetraacetate, Sodium feredetate, Ethylenediaminetetra-acetic acid iron (III) monosodium salt
CAS: 15708-41-5; EC No: 239-802-2; MDL No: MFCD00078215
Powder; Molecular Formula: C10H12FeN2NaO8; MW: 367.05
- Powder
Formula:
C₁₀H₁₂FeN₂NaO₈ MW: 367.05 g/mol |
MDL Number:
MFCD00078215 CAS Number: 15708-41-5 EINECS: 239-802-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...