2,2-Diphenylglycine ≥98%

Supplier: BeanTown Chemical

Synonyms: alpha,alpha-Diphenylglycine, Diphenylglycin, 2-Amino-2,2-diphenylacetic acid

218575-25G 218575-5G
BT218575-25GEA 401 USD
BT218575-25G BT218575-5G
2,2-Diphenylglycine ≥98%
2,2-Diphenylglycine

CAS: 3060-50-2; EC No: 221-299-6; MDL No: MFCD00008048 Solid; Molecular Formula: C14H13NO2; MW: 227.26 Melting Point: 245-247° (decomposes)


  • Solid

Formula: C₁₄H₁₃NO₂
MW: 227.26 g/mol
Melting Pt: 245…247 °C (dec.)
MDL Number: MFCD00008048
CAS Number: 3060-50-2
EINECS: 221-299-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR