Benzyloxyacetaldehyde diethyl acetal ≥98%

Supplier: BeanTown Chemical

218565-10G 218565-250G 218565-50G
BT218565-10GEA 89 USD
BT218565-10G BT218565-250G BT218565-50G
Benzyloxyacetaldehyde diethyl acetal ≥98%
Benzyloxyacetaldehyde diethyl acetal

CAS: 42783-78-8; MDL No: MFCD00051786 Liquid; Molecular Formula: C13H20O3; MW: 224.30 Boiling Point: 99-100°/0.6 mmHg; Flash point: 79°C (174°F) Density (g/mL): 0.987; Refractive Index: 1.478


  • Refractive index: 1.478
  • Liquid

Formula: C₁₃H₂₀O₃
Boiling Pt: 99…100 °C/0.6 mmHg
Density: 0.987 g/mL
Flash Pt: 79 °C (174 °F)
MDL Number: MFCD00051786
CAS Number: 42783-78-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR