(4-Bromophenyl)acetone ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Bromophenylacetone

218560-25G 218560-5G
BT218560-25GEA 378 USD
BT218560-25G BT218560-5G
(4-Bromophenyl)acetone ≥98%
(4-Bromophenyl)acetone

CAS: 6186-22-7; MDL No: MFCD00210401 Liquid; Molecular Formula: C9H9BrO; MW: 213.07


  • Liquid

Formula: C₉H₉BrO
MW: 213.07 g/mol
MDL Number: MFCD00210401
CAS Number: 6186-22-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR