Nickel(II) acetylacetonate 95

Supplier: BeanTown Chemical

Synonyms: Bis(2,4-pentanedionato-O,O')nickel (II), Nickel (II) bis(2,4-pentanedionato-O,O'), Nickel acetylacetonate, Nickel (II) 2,4-pentanedionate

218555-100G 218555-25G 218555-5G
BT218555-100GEA 101.1 USD
BT218555-100G BT218555-25G BT218555-5G
Nickel(II) acetylacetonate 95
Nickel(II) acetylacetonate

CAS: 3264-82-2; EC No: 221-875-7; MDL No: MFCD00000024; RTECS: SA2100000 Powder; Linear Formula: Ni(C5H7O2)2; Molecular Formula: C10H14NiO4 ; MW: 256.91 Melting Point: 230° (decomposes) Hygroscopic


  • RTECS: SA2100000
  • Hygroscopic
  • Powder

Formula: C₁₀H₁₄NiO₄
MW: 256.91 g/mol
Boiling Pt: 220
Melting Pt: 208 (dec.)
MDL Number: MFCD00000024
CAS Number: 3264-82-2
EINECS: 221-875-7
Merck Index: 12,06584

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR