4-Benzyl-L-aspartate ≥98%

Supplier: BeanTown Chemical

Synonyms: L-Aspartic acid beta-benzyl ester, β-Benzyl L-aspartate, H-Asp(obzl)-OH, beta-Benzyl L-aspartate, L-Aspartic acid 4-benzyl ester, L-Aspartic Acid 4-Benzyl Ester, beta-Benzyl L-Aspartate, H-Asp(OBzl)-OH, 4-Benzyl L-Aspartate, H-Asp(OBn)-OH, L-Aspartic Acid beta-Benzyl Ester, L-Aspartic acid β-benzyl ester

218550-1G 218550-25G 218550-5G
BT218550-1GEA 21 USD
BT218550-1G BT218550-25G BT218550-5G
4-Benzyl-L-aspartate ≥98%
4-Benzyl-L-aspartate

CAS: 2177-63-1; EC No: 218-541-8; MDL No: MFCD00037208 Powder; Molecular Formula: C11H13NO4; MW: 223.23


  • Powder

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD00037208
CAS Number: 2177-63-1
EINECS: 218-541-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR