Sodium salicylate ≥99%

Supplier: BeanTown Chemical

Synonyms: Sodium 2-hydroxybenzoate, 2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt

218535-1KG 218535-500G
BT218535-1KGEA 170.94 USD
BT218535-1KG BT218535-500G
Sodium salicylate ≥99%
Sodium salicylate

CAS: 54-21-7; EC No: 200-198-0; MDL No: MFCD00002440; RTECS: VO5075000 Crystalline; Linear Formula: HOC6H4COONa; Molecular Formula: C7H5NaO3; MW: 160.11 Melting Point: 300° Density (g/mL): 0.35 Light Sensitive


  • RTECS: VO5075000
  • Light Sensitive
  • Crystalline

Formula: HOC₆H₄COONa
MW: 160.10 g/mol
Melting Pt: >300 °C
Density: 0.35 g/mL
MDL Number: MFCD00002440
CAS Number: 54-21-7
EINECS: 200-198-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR