1-tert-Butyl-3-ethylbenzene ≥98%

Supplier: BeanTown Chemical

218525-25G 218525-5G
BT218525-25GEA 296 USD
BT218525-25G BT218525-5G
1-tert-Butyl-3-ethylbenzene ≥98%
1-tert-Butyl-3-ethylbenzene

CAS: 14411-56-4; MDL No: MFCD00060845 Liquid; Molecular Formula: C12H18; MW: 162.27


  • Liquid

Formula: C₁₂H₁₈
MW: 162.27 g/mol
MDL Number: MFCD00060845
CAS Number: 14411-56-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR