4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methylsulfonylamino)pyrimidine-5-methanol

Supplier: BeanTown Chemical

Synonyms: 4-(4-Fluorophenyl)-6-isopropyl-2-[(N-methyl-N-methylsufonyl)amino]pyrimidine-5-yl-methanol

218490-10G 218490-2G
BT218490-10GEA 234 USD
BT218490-10G BT218490-2G
4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methylsulfonylamino)pyrimidine-5-methanol
4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methylsulfonylamino)pyrimidine-5-methanol

CAS: 147118-36-3; MDL No: MFCD08063917 Solid; Molecular Formula: C16H20FN3O3S; MW: 353.41


  • Solid

Formula: C₁₆H₂₀FN₃O₃S
MW: 353.412 g/mol
MDL Number: MFCD08063917
CAS Number: 147118-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR