6-Methoxy-2-naphthaldehyde ≥99%

Supplier: BeanTown Chemical

218445-100G 218445-25G 218445-5G
BT218445-100GEA 429 USD
BT218445-100G BT218445-25G BT218445-5G
6-Methoxy-2-naphthaldehyde ≥99%
6-Methoxy-2-naphthaldehyde

CAS: 3453-33-6; EC No: 222-377-2; MDL No: MFCD00081152 Crystalline; Molecular Formula: C12H10O2; MW: 186.21 Melting Point: 81-84° Air Sensitive


  • Air Sensitive
  • Crystalline

Formula: C₁₂H₁₀O₂
Melting Pt: 81…84 °C
MDL Number: MFCD00081152
CAS Number: 3453-33-6
EINECS: 222-377-2

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR