trans-Cinnamic acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
Cinnamic acid (only trans), (2E)-2-Phenyl-2-propenoic acid, trans-Cinnamic acid
218415-100G
218415-2.5KG
218415-500G
BT218415-100GEA
31.2
USD
BT218415-100G
BT218415-2.5KG
BT218415-500G
trans-Cinnamic acid ≥99%
trans-Cinnamic acid
CAS: 140-10-3; EC No: 205-398-1; MDL No: MFCD00004369; RTECS: GD7850000
Crystalline; Linear Formula: C6H5CH=CHCOOH; Molecular Formula: C9H8O2; MW: 148.16
Melting Point: 132-135°; Boiling Point: 300°; Flash point: 100°C (212°F)
Density (g/mL): 1.248
- RTECS: GD7850000
- Crystalline
Formula:
C₉H₈O₂ MW: 148.16 g/mol Boiling Pt: 300 Melting Pt: 132…135 °C Density: 1.248 g/mL Flash Pt: 100 °C (212 °F) |
MDL Number:
MFCD00004369 CAS Number: 140-10-3 EINECS: 205-398-1 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...