Tin(II) pyrophosphate

Supplier: BeanTown Chemical

Synonyms: Stannous pyrophosphate

218385-250G 218385-50G
BT218385-250GEA 98 USD
BT218385-250G BT218385-50G
Tin(II) pyrophosphate
Tin(II) pyrophosphate

CAS: 15578-26-4; EC No: 239-635-5; MDL No: MFCD00049544 Powder; Linear Formula: Sn2P2O7; MW: 411.32 Melting Point: 400° (decomposes) Density (g/mL): 4.009


  • Powder

Formula: O₇P₂Sn₂
MW: 411.361 g/mol
Melting Pt: 400 °C (dec.)
Density: 4.009 g/mL
MDL Number: MFCD00049544
CAS Number: 15578-26-4
EINECS: 239-635-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR