Polypropylene glycol, M.W. 1,000

Supplier: BeanTown Chemical

Synonyms: Poly(propylene glycol), Poly(propylene oxide), PPG

218380-250G 218380-50G
BT218380-250GEA 64.94 USD
BT218380-250G BT218380-50G
Polypropylene glycol, M.W. 1,000
Polypropylene glycol

CAS: 25322-69-4; MDL No: MFCD01779683 Liquid; Linear Formula: H[OCH(CH3)CH2]nOH; MW ≈1,000 Density (g/mL): 1.004


  • Liquid

Formula: H(OCH₂CH₂CH₂)nOH
Density: 1.004 g/mL
MDL Number: MFCD00084446
CAS Number: 25322-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR