[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Dichloro[1,3-bis(diphenylphosphino)propane]palladium(II)
218370-1G
218370-250MG
BT218370-1GEA
255
USD
BT218370-1G
BT218370-250MG
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride ≥98%
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride
CAS: 59831-02-6; MDL No: MFCD03844773
Solid; Molecular Formula: C27H26Cl2P2Pd; MW: 589.77
Air Sensitive
- Air Sensitive
- Solid
Formula:
C₂₇H₂₆Cl₂P₂Pd MW: 589.77 g/mol |
MDL Number:
MFCD03844773 CAS Number: 59831-02-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...