4-(Methylsulfonyl)phenylacetic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-(4-(Methylsulphonyl)phenyl)acetic acid

218360-25G 218360-5G
BT218360-25GEA 176 USD
BT218360-25G BT218360-5G
4-(Methylsulfonyl)phenylacetic acid ≥97%
4-(Methylsulfonyl)phenylacetic acid

CAS: 90536-66-6; MDL No: MFCD00216495 Solid; Molecular Formula: C9H10O4S; MW: 214.24 Melting Point: 136-140°


  • Solid

Formula: C₉H₁₀O₄S
MW: 214.24 g/mol
Melting Pt: 136…140 °C
MDL Number: MFCD00216495
CAS Number: 90536-66-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR