(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine ≥96%
Supplier: BeanTown Chemical
Synonyms:
tert-Butyl (2S,4R)-4-hydroxy-2-hydroxymethylpyrrolidine-1-carboxylate, N-Boc-trans-4-hydroxy-L-prolinol, (2S,4R)-1-Boc-4-hydroxy-2-(hydroxymethyl)pyrrolidine, Boc-Hyp-OL
218345-1G
218345-5G
BT218345-1GEA
136
USD
BT218345-1G
BT218345-5G
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine ≥96%
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine
CAS: 61478-26-0; MDL No: MFCD02094386
Powder; Molecular Formula: C10H19NO4; MW: 217.26
Melting Point: 81-86°
Optical Rotation: [α]20/D -47°, c = 1 in chloroform
Air Sensitive
- Optical rotation: [α]20/D -47°, c = 1 in chloroform
- Air Sensitive
- Powder
Formula:
C₁₀H₁₉NO₄ MW: 217.27 g/mol Melting Pt: 81…86 °C |
MDL Number:
MFCD02094386 CAS Number: 61478-26-0 |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...