Tricyclohexylphosphine ≥96%

Supplier: BeanTown Chemical

218310-1G 218310-25G 218310-5G
BT218310-1GEA 34.61 USD
BT218310-1G BT218310-25G BT218310-5G
Tricyclohexylphosphine ≥96%
Tricyclohexylphosphine

CAS: 2622-14-2; EC No: 220-069-2; MDL No: MFCD00003853 Crystalline/Powder; Linear Formula: (C6H11)3P ; MW: 280.42 Melting Point: 81-83° Air Sensitive


  • Air Sensitive
  • Crystalline/Powder

Formula: C₁₈H₃₃P
Melting Pt: 81…83 °C
MDL Number: MFCD00003853
CAS Number: 2622-14-2
EINECS: 220-069-2

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR