(1-Tetradecyl)triphenylphosphonium bromide ≥97%

Supplier: BeanTown Chemical

Synonyms: Triphenyl(tetradecyl)phosphonium bromide, Tetradecyltriphenylphosphonium bromide

218290-100G 218290-25G
BT218290-100GEA 282 USD
BT218290-100G BT218290-25G
(1-Tetradecyl)triphenylphosphonium bromide ≥97%
(1-Tetradecyl)triphenylphosphonium bromide

CAS: 25791-20-2; EC No: 247-267-1; MDL No: MFCD00051857 Powder; Linear Formula: CH3(CH2)12P(Br)(C6H5)3; Molecular Formula: C31H42BrP; MW: 539.59 Melting Point: 87-89°


  • Powder

Formula: H₃C(CH₂)₁₃P(C₆H₅)₃Br
MW: 539.58 g/mol
Melting Pt: 87…89 °C
MDL Number: MFCD00051857
CAS Number: 25791-20-2
EINECS: 247-267-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR