N,O-Dimethylhydroxylamine hydrochloride ≥98%

Supplier: BeanTown Chemical

218280-100G 218280-25G 218280-5G
BT218280-100GEA 297 USD
BT218280-100G BT218280-25G BT218280-5G
N,O-Dimethylhydroxylamine hydrochloride ≥98%
N,O-Dimethylhydroxylamine hydrochloride

CAS: 6638-79-5; EC No: 229-642-1; MDL No: MFCD00012485 Crystalline/Powder; Linear Formula: CH3ONHCH3·HCl; Molecular Formula: C2H8ClNO; MW: 97.55 Melting Point: 112-115° Hygroscopic


  • Hygroscopic
  • Crystalline/Powder

Formula: C₂H₈ClNO
MW: 97.55 g/mol
Melting Pt: 112…115 °C
MDL Number: MFCD00012485
CAS Number: 6638-79-5
EINECS: 229-642-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR