2-Methylnicotinic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Methylpyridine-3-carboxylic acid, 2-Methyl-3-pyridinecarboxylic acid
218245-1G
218245-5G
BT218245-1GEA
67
USD
BT218245-1G
BT218245-5G
2-Methylnicotinic acid ≥98%
2-Methylnicotinic acid
CAS: 3222-56-8; MDL No: MFCD00013449
Powder; Molecular Formula: C7H7NO2; MW: 137.14
Melting Point: 228-230° (decomposes)
- Powder
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Melting Pt: 228…230 °C (dec.) |
MDL Number:
MFCD00013449 CAS Number: 3222-56-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...