Molybdenum carbide ≥99.5%
Supplier: BeanTown Chemical
218205-100G
218205-500G
BT218205-100GEA
80.88
USD
BT218205-100G
BT218205-500G
Molybdenum carbide ≥99.5%
Molybdenum carbide
CAS: 12069-89-5; EC No: 235-115-7; MDL No: MFCD00014218
-325 Mesh Powder; Linear Formula: Mo2C; MW: 203.89
Melting Point: 2687°
- -325 Mesh Powder
Formula:
CMo₂ MW: 203.89 g/mol Melting Pt: 2687 °C |
MDL Number:
MFCD00014218 CAS Number: 12069-89-5 EINECS: 235-115-7 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...