Imidazole-4-carboxaldehyde ≥97%

Supplier: BeanTown Chemical

Synonyms: 1H-Imidazole-5-carboxaldehyde, 4-Formylimidazole, Imidazole-4-carbaldehyde

218190-1G 218190-25G 218190-5G
BT218190-1GEA 34 USD
BT218190-1G BT218190-25G BT218190-5G
Imidazole-4-carboxaldehyde ≥97%
Imidazole-4-carboxaldehyde

CAS: 3034-50-2; EC No: 221-227-3; MDL No: MFCD00173726 Solid; Molecular Formula: C4H4N2O; MW: 96.09 Melting Point: 174-177° Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₄H₄N₂O
MW: 96.09 g/mol
Melting Pt: 174…177 °C
MDL Number: MFCD00173726
CAS Number: 3034-50-2
EINECS: 221-227-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR