Starch, soluble

Supplier: BeanTown Chemical

Synonyms: Starch, Vitex® iodine indicator, Amylodextrin

218180-100G 218180-500G
BT218180-100GEA 38 USD
BT218180-100G BT218180-500G
Starch, soluble
Starch, soluble

CAS: 9005-84-9; EC No: 232-686-4; MDL No: MFCD00082026 Powder; Molecular Formula: C12H22O11; MW: 342.30 Melting Point: 256-258° (decomposes)


  • Powder

Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Melting Pt: 256…258 °C (dec.)
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR