trans-1,2-Cyclohexanedicarboxylic acid ≥98%
Supplier: BeanTown Chemical
218170-25G
218170-5G
BT218170-25GEA
189.88
USD
BT218170-25G
BT218170-5G
trans-1,2-Cyclohexanedicarboxylic acid ≥98%
trans-1,2-Cyclohexanedicarboxylic acid
CAS: 2305-32-0; EC No: 218-975-8; MDL No: MFCD00001464; RTECS: GU9065000
Solid; Linear Formula: C6H10(COOH)2; Molecular Formula: C8H12O4; MW: 172.18
Melting Point: 228-230°
- RTECS: GU9065000
- Solid
Formula:
C₈H₁₂O₄ Melting Pt: 228…230 °C |
MDL Number:
MFCD00001464 CAS Number: 2305-32-0 EINECS: 218-975-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...