Sodium bromide ≥99.99%

Supplier: BeanTown Chemical

218120-25G 218120-5G
BT218120-25GEA 192.99 USD
BT218120-25G BT218120-5G
Sodium bromide ≥99.99%
Sodium bromide

CAS: 7647-15-6; EC No: 231-599-9; MDL No: MFCD00003475; RTECS: VZ3150000 Crystalline; Linear Formula: NaBr; MW: 102.89 Melting Point: 755°; Boiling Point: 1393° Density (g/mL): 3.203; Refractive Index: 1.641 Hygroscopic


  • RTECS: VZ3150000
  • Refractive index: 1.641
  • Hygroscopic
  • Crystalline

Formula: NaBr
MW: 102.89 g/mol
Boiling Pt: 1396
Melting Pt: 753
Density: 3.203 g/mL
MDL Number: MFCD00003475
CAS Number: 7647-15-6
EINECS: 231-599-9
Merck Index: 13,08666

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR