(R)-(+)-1,2,3,4-Tetrahydro-1-naphthoic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: (R)-1,2,3,4-Tetrahydronaphthoic acid

218115-1G 218115-250MG 218115-5G
BT218115-1GEA 200 USD
BT218115-1G BT218115-250MG BT218115-5G
(R)-(+)-1,2,3,4-Tetrahydro-1-naphthoic acid ≥98%
(R)-(+)-1,2,3,4-Tetrahydro-1-naphthoic acid

CAS: 23357-47-3; MDL No: MFCD07368364 Crystalline/Powder; Molecular Formula: C11H12O2 ; MW: 176.21 Boiling Point: 54-57°


  • Crystalline/Powder

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Boiling Pt: 54…57 °C
MDL Number: MFCD07368364
CAS Number: 23357-47-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR