Phenyl phosphate disodium salt, hydrate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Phosphoric acid phenyl ester disodium salt, Phosphoric acid monophenyl ester disodium salt
218100-100G
218100-25G
BT218100-100GEA
220
USD
BT218100-100G
BT218100-25G
Phenyl phosphate disodium salt, hydrate ≥98%
Phenyl phosphate disodium salt
CAS: 3279-54-7; EC No: 221-917-4; MDL No: MFCD00002133
Crystalline/Powder; Linear Formula: C6H5PO4Na2·xH2O; MW: 218.06 (anhydrous)
Hygroscopic
- Crystalline/Powder
- Hygroscopic
Formula:
C₆H₅Na₂O₄P·xH₂O MW: 218.06 g/mol (anhy) |
MDL Number:
MFCD00002133 CAS Number: 3279-54-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...