Glycine hydrochloride (H-Gly-OH.HCl) ≥98%
Supplier: BeanTown Chemical
Synonyms:
H-Gly-OH·HCl, Glycine hydrochloride
218010-100G
218010-2.5KG
218010-500G
BT218010-100GEA
44.31
USD
BT218010-100G
BT218010-2.5KG
BT218010-500G
Glycine hydrochloride (H-Gly-OH.HCl) ≥98%
Glycine hydrochloride (H-Gly-OH.HCl)
CAS: 6000-43-7; EC No: 227-841-8; MDL No: MFCD00012872; RTECS: MC0560000
Solid; Linear Formula: NH2CH2CO2H·HCl; Molecular Formula: C2H5NO2·HCl; MW: 111.53
Melting Point: 176-180°
Hygroscopic
- RTECS: MC0560000
- Hygroscopic
- Solid
Formula:
C₂H₆ClNO₂ MW: 111.53 g/mol Melting Pt: 176…180 °C |
MDL Number:
MFCD00012872 CAS Number: 6000-43-7 EINECS: 227-841-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...