3,5-Dihydroxybenzaldehyde ≥97%

Supplier: BeanTown Chemical

218000-1G 218000-25G 218000-5G
BT218000-1GEA 80 USD
BT218000-1G BT218000-25G BT218000-5G
3,5-Dihydroxybenzaldehyde ≥97%
3,5-Dihydroxybenzaldehyde

CAS: 26153-38-8; EC No: 247-479-4; MDL No: MFCD00016611 Powder; Molecular Formula: C7H6O3; MW: 138.12 Melting Point: 153-158° Air Sensitive


  • Air Sensitive
  • Powder

Formula: C₇H₆O₃
MW: 138.12 g/mol
Melting Pt: 153…158 °C
MDL Number: MFCD00016611
CAS Number: 26153-38-8
EINECS: 247-479-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR