2-Methyl-1[4-(trifluoromethoxy)phenyl]propan-1-one

Supplier: BeanTown Chemical

Synonyms: 2-Methyl-1(4-trifluoromethoxyphenyl)propan-1-one

217975-1G 217975-5G
BT217975-1GEA 208 USD
BT217975-1G BT217975-5G
2-Methyl-1[4-(trifluoromethoxy)phenyl]propan-1-one
2-Methyl-1[4-(trifluoromethoxy)phenyl]propan-1-one

CAS: 56425-84-4; EC No: 260-171-4; MDL No: MFCD06412566 Liquid/Low Melting Solid; Molecular Formula: C11H11F3O2; MW: 232.20 Melting Point: 17°; Boiling Point: 248° Density (g/mL): 1.192


  • Liquid/Low Melting Solid

Formula: C₁₁H₁₁F₃O₂
MW: 232.202 g/mol
Boiling Pt: 248 °C
Melting Pt: 17 °C
Density: 1.192 g/mL
MDL Number: MFCD06412566
CAS Number: 56425-84-4
EINECS: 260-171-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR