Tetraethyl methylenebisphosphonate ≥98%

Supplier: BeanTown Chemical

Synonyms: Tetraethyl methylenediphosphonate, Tetraethyl methylenebis(phosphonate)

217950-100G 217950-25G 217950-5G
BT217950-100GEA 937 USD
BT217950-100G BT217950-25G BT217950-5G
Tetraethyl methylenebisphosphonate ≥98%
Tetraethyl methylenebisphosphonate

CAS: 1660-94-2; EC No: 216-764-5; MDL No: MFCD00039887; RTECS: SZ9150000 Liquid; Linear Formula: CH2[P(O)(OCH2CH3)2]2; Molecular Formula: C9H22O6P2; MW: 288.22 Boiling Point: 171-174°/11 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.162; Refractive Index: 1.442


  • RTECS: SZ9150000
  • Refractive index: 1.442
  • Liquid

Formula: C₉H₂₂O₆P₂
MW: 288.22 g/mol
Boiling Pt: 171…174 °C/11 mmHg
Density: 1.162 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00039887
CAS Number: 1660-94-2
EINECS: 216-764-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR