Stearic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
Octadecanoic acid, Octadecylic acid
217930-100G
217930-25G
217930-5G
BT217930-100GEA
317.42
USD
BT217930-100G
BT217930-25G
BT217930-5G
Stearic acid ≥98%
Stearic acid
CAS: 57-11-4; EC No: 200-313-4; MDL No: MFCD00002752; RTECS: WI2800000
Solid; Linear Formula: CH3(CH2)16COOH; Molecular Formula: C18H36O2; MW: 284.48
Melting Point: 67-72°; Boiling Point: 361°; Flash point:
- RTECS: WI2800000
- Solid
Formula:
C₁₈H₃₆O₂ MW: 284.48 g/mol Boiling Pt: 361 Melting Pt: 67 Density: 0.845 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00002752 CAS Number: 57-11-4 EINECS: 200-313-4 Merck Index: 13,08882 |
|
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...