Iron(III) hydroxide, α-phase

Supplier: BeanTown Chemical

217885-25G 217885-5G
BT217885-25GEA 648 USD
BT217885-25G BT217885-5G
Iron(III) hydroxide, α-phase
Iron(III) hydroxide, alpha-phase

CAS: 20344-49-4; EC No: 243-746-4; MDL No: MFCD00064782 Powder; Linear Formula: FeO(OH); MW: 88.85 Density (g/mL): 3.4-3.9


  • Powder

Formula: FeO(OH)
Density: 3.4…3.9 g/mL
MDL Number: MFCD00064782
CAS Number: 20344-49-4
EINECS: 243-746-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR