1-Boc-3-fluoro-4-pyrrolidinone
Supplier: BeanTown Chemical
Synonyms:
tert-Butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate, 4-Fluoropyrrolidin-3-one, N-BOC protected
217860-1G
217860-250MG
BT217860-1GEA
1159
USD
BT217860-1G
BT217860-250MG
1-Boc-3-fluoro-4-pyrrolidinone
1-Boc-3-fluoro-4-pyrrolidinone
CAS: 845894-03-3; MDL No: MFCD08274459
Solid; Molecular Formula: C9H14FNO3; MW: 203.22
- Solid
Formula:
C₉H₁₄FNO₃ MW: 203.213 g/mol |
MDL Number:
MFCD08274459 CAS Number: 845894-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...