Molecular sieve A5 (0.5 nm, 5 Å), beads 3-5 mm (0.12-0.20 in)
Supplier: BeanTown Chemical
217855-1KG
217855-250G
BT217855-1KGEA
80.23
USD
BT217855-1KG
BT217855-250G
Molecular sieve A5 (0.5 nm, 5 Å), beads 3-5 mm (0.12-0.20 in)
Molecular sieve A5 (0.5 nm, 5 Å)
CAS: 69912-79-4; EC No: 215-684-8; MDL No: MFCD00131613; RTECS: ZG6800000
Beads
Density (g/mL): 1.44
Hygroscopic
- RTECS: ZG6800000
- Hygroscopic
- Beads
Density:
1.44 g/mL |
MDL Number:
MFCD00131613 CAS Number: 69912-79-4 EINECS: 215-684-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...