2,4,6-Triisopropylbenzenesulfonyl hydrazide
Supplier: BeanTown Chemical
Synonyms:
2,4,6-Triisopropylbenzenesulphonohydrazide
217845-2G
217845-500MG
BT217845-2GEA
117
USD
BT217845-2G
BT217845-500MG
2,4,6-Triisopropylbenzenesulfonyl hydrazide
2,4,6-Triisopropylbenzenesulfonyl hydrazide
CAS: 39085-59-1; EC No: 254-282-7; MDL No: MFCD00014750
Crystalline; Molecular Formula: C15H26N2O2S; MW: 298.44
Melting Point: 110-112° (decomposes)
Moisture Sensitive
- Moisture Sensitive
- Crystalline
Formula:
C₁₅H₂₆N₂O₂S Melting Pt: 110…112 °C (dec.) |
MDL Number:
MFCD00014750 CAS Number: 39085-59-1 EINECS: 254-282-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...