1-Aminopyrene ≥97%
Supplier: BeanTown Chemical
Synonyms:
Pyren-1-ylamine, Pyren-1-amine
217840-2G
217840-500MG
BT217840-2GEA
145
USD
BT217840-2G
BT217840-500MG
1-Aminopyrene ≥97%
1-Aminopyrene
CAS: 1606-67-3; EC No: 216-521-3; MDL No: MFCD00004140
Powder; Molecular Formula: C16H11N; MW: 217.27
Melting Point: 115-117°
- Powder
Formula:
C₁₆H₁₁N MW: 217.27 g/mol Melting Pt: 115…117 °C |
MDL Number:
MFCD00004140 CAS Number: 1606-67-3 EINECS: 216-521-3 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...