Di-n-octyl phthalate ≥98%

Supplier: BeanTown Chemical

217815-100ML 217815-25ML 217815-500ML
BT217815-100MLEA 123 USD
BT217815-100ML BT217815-25ML BT217815-500ML
Di-n-octyl phthalate ≥98%
Di-n-octyl phthalate

CAS: 117-84-0; EC No: 204-214-7; MDL No: MFCD00015292; RTECS: TI1925000 Liquid; Molecular Formula: C24H38O4; MW: 390.56 Flash point: 109°C (228°F) Density (g/mL): 0.98


  • RTECS: TI1925000
  • Liquid

Formula: C₂₄H₃₈O₄
Density: 0.98 g/mL
Flash Pt: 109 °C (228 °F)
MDL Number: MFCD00015292
CAS Number: 117-84-0
EINECS: 204-214-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR