Strontium chloride, anhydrous ≥95%
Supplier: BeanTown Chemical
217800-2KG
217800-500G
BT217800-2KGEA
160
USD
BT217800-2KG
BT217800-500G
Strontium chloride, anhydrous ≥95%
Strontium chloride
CAS: 10476-85-4; EC No: 233-971-6; MDL No: MFCD00011249
Powder; Linear Formula: SrCl2; MW: 158.53
Melting Point: 873°; Boiling Point: 1250°
Density (g/mL): 3.052; Refractive Index: 1.65
Hygroscopic
- Refractive index: 1.65
- Hygroscopic
- Powder
Formula:
Cl₂Sr Boiling Pt: 1250 °C Melting Pt: 873 °C Density: 3.052 g/mL |
MDL Number:
MFCD00011249 CAS Number: 10476-85-4 EINECS: 233-971-6 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...