(S)-β-Phenylalaninol ≥97%
Supplier: BeanTown Chemical
Synonyms:
(S)-3-Amino-3-phenyl-1-propanol, (S)-3-Amino-3-phenylpropan-1-ol
217765-1G
217765-5G
BT217765-1GEA
216
USD
BT217765-1G
BT217765-5G
(S)-β-Phenylalaninol ≥97%
(S)-β-Phenylalaninol
CAS: 82769-76-4; MDL No: MFCD01311768
Solid; Molecular Formula: C9H13NO; MW: 151.21
- Solid
Formula:
C₉H₁₃NO MW: 151.2056 g/mol |
MDL Number:
MFCD01311768 CAS Number: 82769-76-4 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...