Sodium dihydrogen citrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Sodium 3,4-dicarboxy-3-hydroxybutanoate, Sodium citrate monobasic, mono-Sodium citrate, Citric acid monosodium salt, Monosodium Citrate
217710-100G
217710-2KG
217710-500G
BT217710-100GEA
25
USD
BT217710-100G
BT217710-2KG
BT217710-500G
Sodium dihydrogen citrate ≥99%
Sodium dihydrogen citrate
CAS: 18996-35-5; EC No: 242-734-6; MDL No: MFCD00013067; RTECS: GE7950000
Crystalline; Linear Formula: HOC(COONa)(CH2COOH)2; Molecular Formula: C6H7NaO7; MW: 214.11
Melting Point: 208°
Hygroscopic
- RTECS: GE7950000
- Hygroscopic
- Crystalline
Formula:
C₆H₇NaO₇ MW: 214.11 g/mol Melting Pt: 208 °C |
MDL Number:
MFCD00013067 CAS Number: 18996-35-5 EINECS: 242-734-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...