Tropinone ≥99%

Supplier: BeanTown Chemical

Synonyms: 8-Methyl-8-azabicyclo[3.2.1]octan-3-one

217700-100G 217700-25G 217700-5G
BT217700-100GEA 519 USD
BT217700-100G BT217700-25G BT217700-5G
Tropinone ≥99%
Tropinone

CAS: 532-24-1; EC No: 208-530-6; MDL No: MFCD00005549 Crystalline; Molecular Formula: C8H13NO; MW: 139.20 Melting Point: 40-44°; Boiling Point: 227°; Flash point: 90°C (194°F)


  • Crystalline

Formula: C₈H₁₃NO
MW: 139.2 g/mol
Boiling Pt: 227 °C
Melting Pt: 40…44 °C
Flash Pt: 90 °C (194 °F)
MDL Number: MFCD00005549
CAS Number: 532-24-1
EINECS: 208-530-6

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR