Salicylic acid ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: 2-Hydroxybenzoic acid, o-Hydroxybenzoic acid

217650-100G 217650-2.5KG 217650-500G
BT217650-100GEA 33.18 USD
BT217650-100G BT217650-2.5KG BT217650-500G
Salicylic acid ≥99.0% ACS
Salicylic acid

CAS: 69-72-7; EC No: 200-712-3; MDL No: MFCD00002439; RTECS: VO0525000 Crystalline/Powder; Linear Formula: HOC6H4COOH; Molecular Formula: C7H6O3; MW: 138.12 Melting Point: 158-161°; Boiling Point: 211°/20 mmHg; Flash point: 157°C (315°F) Density (g/mL): 1.443 Light Sensitive


  • RTECS: VO0525000
  • Light Sensitive
  • Crystalline/Powder

Formula: (HO)C₆H₄CO₂H
MW: 138.12 g/mol
Boiling Pt: 336
Melting Pt: 158…161 °C
Density: 1.443 g/mL
Flash Pt: 157 °C (315 °F)
MDL Number: MFCD00002439
CAS Number: 69-72-7
EINECS: 200-712-3
Merck Index: 13,08411
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af1f695fa92656a135b5e1742f1cca28f4d255fb&vid=8f3aff6fb9a70e5dd194b8bc46019af74e48e90a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.
Melting range 158.0-161.0°C
Residue after ignition ≤0.01%
Chloride (Cl) ≤0.001%
Sulfate (SO4) ≤0.003%
Heavy metals (as Pb) ≤5ppm
Iron (Fe) ≤2ppm
Substance darkened by sulfuric acid Passes test
Appearance White, fine crystalline powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR