Palladium(II) oxide, anhydrous ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
217635-1G
217635-5G
BT217635-1GEA
245
USD
BT217635-1G
BT217635-5G
Palladium(II) oxide, anhydrous ≥99.9% (trace metals basis)
Palladium(II) oxide
CAS: 1314-08-5; EC No: 215-218-3; MDL No: MFCD00011172; RTECS: RT3592900
-22 Mesh Powder; Linear Formula: PdO; MW: 122.42
Melting Point: 870°
Density (g/mL): 8.7
- RTECS: RT3592900
- -22 Mesh Powder
Formula:
OPd MW: 122.42 g/mol Melting Pt: 870 °C Density: 8.7 g/mL |
MDL Number:
MFCD00011172 CAS Number: 1314-08-5 EINECS: 215-218-3 |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...